Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK5RAP3, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300870AM

 View more versions of this product

Catalog No. 50-155-8664


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

This gene encodes a protein that has been reported to function in signaling pathways governing transcriptional regulation and cell cycle progression. It may play a role in tumorigenesis and metastasis. A pseudogene of this gene is located on the long arm of chromosome 20. Alternative splicing results in multiple transcript variants that encode different isoforms. [provided by RefSeq, May 2013].
Specifications

Specifications

CDK5RAP3
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q96JB5
IC53-2
Rabbit
IgG
100 μl
4° C
Primary
80279
Western Blot
Unconjugated
CDK5RAP3
liquid
CDK5RAP3
Between 275 and 325
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit