Learn More
Aviva Systems Biology Corp CDK7 Antibody - middle region (P100995_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDK7. Alternate names: CAK1, CDKN7, MO15, STK1, p39MO15, HCAKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-62822
Add to Cart
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'