Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
$562.00 - $1916.00
Specifications
Gene Alias | CDK7/Cyclin H/MNAT1 |
---|---|
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Description
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle.Specifications
CDK7/Cyclin H/MNAT1 | |
Human | |
His Tag | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title