Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK7 Mouse anti-Human, Clone: E3E, Abnova™

Mouse monoclonal antibody raised against human CDK7.

Manufacturer:  Abnova Corporation MAB12702

Catalog No. 89-933-922


Add to cart

Description

Description

The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle. [provided by RefSeq]

Specifications

Specifications

CDK7
E3E
Mouse monoclonal antibody raised against human CDK7.
In HEPES, 0.15M NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide).
CAK1/CDKN7/MO15/STK1/p39MO15
Mouse
IgG2a, kappa
50 uL
Store at -20°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Purified
CDK7
Recombinant protein corresponding to amino acids 136-346 human CDK7.
Ammonium sulfate precipitation
RUO
Primary
1022
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit