Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK8/Cyclin C Recombinant Human Protein Non-distribution product as customer accommodation.

CDK8/Cyclin C Recombinant Human Protein

$434.00

Specifications

Accession Number NP_001251, NP_005181
Assays The ability of CDK8/cyclin C to bind the ATP competitor kinase tracer 236 (PV5592) was demonstrated for this lot of CDK8/cyclin C in the LanthaScreen™ Eu kinase binding assay. Increasing amounts of kinase tracer 236 (PV5592) were incubated with 5nM CDK8/cyclin C and 2nM Eu-anti-HIS antibody (PV5596) in kinase buffer A (PV3189) with and without the presence of 10μM Staurosporine as the competitor for the kinase tracer
Common Name CDK8/cyclin C Recombinant Human Protein
Concentration 0.34mg/mL
Expression System Baculovirus
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4402 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV4402
10μg Each for $434.00
Add to cart
 
Description

Description

CDK8 phosphorylates the carboxy-terminal domain of the largest subunit of RNA polymerase II. This protein is coexpressed and copurified with CyclinC.
Specifications

Specifications

NP_001251, NP_005181
CDK8/cyclin C Recombinant Human Protein
Baculovirus
CDK and CHK
1 Each
80% by SDS-PAGE
–80°C
Human
The ability of CDK8/cyclin C to bind the ATP competitor kinase tracer 236 (PV5592) was demonstrated for this lot of CDK8/cyclin C in the LanthaScreen™ Eu kinase binding assay. Increasing amounts of kinase tracer 236 (PV5592) were incubated with 5nM CDK8/cyclin C and 2nM Eu-anti-HIS antibody (PV5596) in kinase buffer A (PV3189) with and without the presence of 10μM Staurosporine as the competitor for the kinase tracer
0.34mg/mL
CDK8/cyclin C, CCNC
CDK8/cyclin C Recombinant Human Protein
CDK8/cyclin C Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit