Learn More
Aviva Systems Biology Corp CDK9 Antibody - N-terminal region (AVARP03004_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP03004_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDK9. Alternate names: C-2k, CDC2L4, PITALRE, TAK, CTK1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-62502