Learn More
CDK9/Cyclin T1 Recombinant Human Protein
$456.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV4131 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV4131
N/A 10μg
Each for $456.00
Add to cart
Description
Description
CDK9 is a serine⁄threonine kinase which when complexed with CyclinT1 forms an active holoenzyme complex. This complex plays a key role in regulating the eukaryotic cell cycle. CDK9 is coexpressed and copurified with CyclinT1.