Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKL1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271389AF405

Catalog No. N271389A405


Add to cart

Description

Description

CDKL1 Monoclonal antibody specifically detects CDKL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CDKL1
OTI1D9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CDKL1
Recombinant protein fragment corresponding to amino acids 152-357 of human CDKL1 (NP_004187) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
8814.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CDC2-related kinase 1, cyclin-dependent kinase-like 1, cyclin-dependent kinase-like 1 (CDC2-related kinase), EC 2.7.11, EC 2.7.11.22, KKIALRE, P42, Protein kinase p42 KKIALRE, serine/threonine protein kinase KKIALRE, Serine/threonine-protein kinase KKIALRE
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit