Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKL4 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297193PCP
Description
CDKL4 Polyclonal antibody specifically detects CDKL4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CDKL4 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
cyclin-dependent kinase-like 4, EC 2.7.11, EC 2.7.11.22 | |
Produced in rabbits immunized with E. coli-derived Human CDKL4 fragment. | |
0.1 mL | |
Protein Kinase | |
344387 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction