Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKL5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179782

 View more versions of this product

Catalog No. NBP179782


Add to cart

Description

Description

CDKL5 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

CDKL5
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
CDKL5
Synthetic peptide directed towards the C terminal of human CDKL5The immunogen for this antibody is CDKL5. Peptide sequence SQASGGSSNIRQEPAPKGRPALQLPDGGCDGRRQRHHSGPQDRRFMLRTT.
115 kDa
100 ul
Protein Kinase
Primary
6792
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 0.2-1 ug/ml
NP_003150
cyclin-dependent kinase-like 5, serine/threonine kinase 9, serine/threonine-protein kinase 9
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%.
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit