Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1A Mouse anti-Human, Clone: DCS-60.2, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14201

Catalog No. 89-034-302


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human CDKN1A.

  • High specificity
  • High reactivity
Specifications

Specifications

CDKN1A
DCS-60.2
Mouse monoclonal antibody raised against full length recombinant human CDKN1A.
In PBS.
Liquid
CDKN1A
Recombinant protein corresponding to full length human CDKN1A.
Protein A purification
RUO
Primary
1026
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the
P38936
CAP20|CDKN1|CIP1|MDA-6|P21|SDI1|WAF1|p21CIP1
Mouse
IgG2a, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody is highly specific to human CDKN1A and shows no cross-reaction with other closely related mitotic inhibitors.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)