Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1C Mouse anti-Human, Clone: KIP2/880, Abnova™

Mouse monoclonal antibody raised against full length recombinant human CDKN1C.

Manufacturer:  Abnova Corporation MAB13250

Catalog No. 89-933-936


Add to cart

Description

Description

The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

CDKN1C
KIP2/880
Mouse monoclonal antibody raised against full length recombinant human CDKN1C.
In 10mM PBS.
BWCR/BWS/KIP2/WBS/p57
Mouse
IgG2b, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This monoclonal antibody shows no cross-reaction with CDKN1B.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CDKN1C
Recombinant protein corresponding to full length human CDKN1C.
Protein A/G purification
RUO
Primary
1028
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit