Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN2A (phospho S152) Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CDKN2A.

Manufacturer:  Abnova Corporation PAB29585

Catalog No. 89-956-742


Add to cart

Description

Description

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, MDM1, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene. [provided by RefSeq]

Specifications

Specifications

CDKN2A (phospho S152)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Liquid
CDKN2A
A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S152 of human CDKN2A.
Affinity chromatography
RUO
Primary
1029
Human
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CDKN2A.
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
ARF|CDK4I|CDKN2|CMM2|INK4|INK4a|MLM|MTS1|TP16|p14|p14ARF|p16|p16INK4|p16INK4a|p19
Rabbit
IgG
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
CDKN2A (phospho S152) polyclonal antibody detects endogenous levels of human CDKN2A only when phosphorylated at serine 152.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit