Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKN3 Mouse anti-Human, DyLight 755, Clone: OTI2E11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270399IR
Description
CDKN3 Monoclonal antibody specifically detects CDKN3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
CDKN3 | |
OTI2E11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
CDKN3 | |
Full length recombinant protein of human CDKN3 (NP_005183) produced in HEK293T cells. | |
Immunogen affinity purified | |
Cell Cycle and Replication, Core ESC Like Genes, Protein Phosphatase, Stem Cell Markers | |
Primary | |
1033.0 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDI1KAP1, CDK2-associated dual specificity phosphatase, CDK2-associated dual-specificity phosphatase, Cdk-associated protein phosphatase, CIP2, cyclin-dependent kinase inhibitor 3, cyclin-dependent kinase interacting protein 2, Cyclin-dependent kinase interactor 1, Cyclin-dependent kinase-interacting protein 2, EC 3.1.3.16, EC 3.1.3.48, FLJ25787, KAPMGC70625, Kinase-associated phosphatase | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title