Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN3 Mouse anti-Human, PE/Cy5.5, Clone: OTI2E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270399PECY55

Catalog No. N270399PEC5


Add to cart

Description

Description

CDKN3 Monoclonal antibody specifically detects CDKN3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CDKN3
OTI2E11
Flow Cytometry
Affinity Purified, Conjugated
CDKN3
Full length recombinant protein of human CDKN3 (NP_005183) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Core ESC Like Genes, Protein Phosphatase, Stem Cell Markers
Primary
1033.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CDI1KAP1, CDK2-associated dual specificity phosphatase, CDK2-associated dual-specificity phosphatase, Cdk-associated protein phosphatase, CIP2, cyclin-dependent kinase inhibitor 3, cyclin-dependent kinase interacting protein 2, Cyclin-dependent kinase interactor 1, Cyclin-dependent kinase-interacting protein 2, EC 3.1.3.16, EC 3.1.3.48, FLJ25787, KAPMGC70625, Kinase-associated phosphatase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit