Learn More
Tocris Bioscience™ CDN 1163 Available on GSA/VA Contract for Federal Government customers only.
SERCA2 allosteric activator
$157.70 - $648.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-691-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5869/10
Each for $157.70
Add to cart
58-695-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5869/50
Each for $648.50
Add to cart
Description
Description
SERCA2 allosteric activator. Increases Ca2+-ATPase activity and Ca2+ uptake by ER microsomes from obese mice. Rescues HEK cells from ER stress-induced cell death. Reduces fasting glucose levels and adipose tissue weight, and increases energy expenditure in ob/ob mice. Also improves glucose tolerance and reverses hepatic steatosis in ob/ob mice. Neuroprotective and improves memory and cognition in a mouse Alzheimer's disease model. Orally bioavailable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.