Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDP 840 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of phosphodiesterase (PDE) 4 (IC50 = 12 nM for native PDE4). Competitively inhibits all PDE4 isoenzymes (IC50 values are 4, 9, 9 and 45 nM for PDE4A, PDE4B, PDE4C and PDE4D human recombinant isoforms expressed in yeast).
Specifications
Quantity | 10 mg |
Formula Weight | 409.95 |
Percent Purity | >98% |
Chemical Name or Material | CDP 840 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction