Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDP/CUTL1 Antibody (CL5278), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CDP/CUTL1 |
---|---|
Clone | CL5278 |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CDP/CUTL1 Monoclonal specifically detects CDP/CUTL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CDP/CUTL1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
CCAAT displacement protein, CDP/Cux, CDPCCAAT displacement protein, COY1, CUT, cut (Drosophila)-like 1 (CCAAT displacement protein), cut-like 1, CCAAT displacement protein (Drosophila), cut-like homeobox 1, FLJ31745, golgi integral membrane protein 6, GOLIM6, homeobox protein cut-like 1, Homeobox protein cux-1, Nbla10317, p100, p110, p200, p75, protein CASP, putative protein product of Nbla10317 | |
CUX1 | |
IgG1 | |
Protein A purified |
CL5278 | |
Monoclonal | |
Purified | |
Human | |
1523 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LDPALKQAPLSQSDITILTPKLLSTSPMPTVSSYPPLAISLKKPSAAPEAGASALPNPPALKKEAQDAPGLDPQGAADCAQGVLRQVKNEVGRSGAWKDHWWS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title