Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDR2 Mouse anti-Human, Clone: 33, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524794

Catalog No. PIMA524794


Add to cart

Description

Description

Does not cross react with mouse Cdr2 and human Cdr3.

Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
Specifications

Specifications

CDR2
Monoclonal
1 mg/mL
500mM tris glycine, 50mM glycine with 0.15M NaCl and 0.05% sodium azide; pH 7.5
Q01850
CDR62, cerebellar degeneration-related protein (62kD), cerebellar degeneration-related protein 2, cerebellar degeneration-related protein 2, 62kDa, Major Yo paraneoplastic antigen, Paraneoplastic cerebellar degeneration-associated antigen, PCD17, Yo, Yo paraneoplastic antigen
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1039
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
33
Unconjugated
CDR2
Liquid
CDR2
N-terminal MBP-tagged full length human CDR2.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit