Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDRT4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548995
Description
Specifications
CDRT4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8N9R6 | |
CDRT4 | |
KLH conjugated synthetic peptide between 50-79 amino acids from the Central region of human CDRT4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CDRT4 | |
CMT1A duplicated region transcript 4 protein, CDRT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284040 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction