Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597574
Description
CDS2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Provides CDP-diacylglycerol, an important precursor for the synthesis of phosphatidylinositol, phosphatidylglycerol, and cardiolipin.
Specifications
CDS2 | |
Polyclonal | |
Unconjugated | |
CDS2 | |
5730450N06Rik; 5730460C18Rik; AI854580; CDP-DAG synthase 2; CDP-DG synthase 2; CDP-DG synthetase 2; CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2; CDP-diacylglycerol synthase 2; CDP-diglyceride diphosphorylase 2; CDP-diglyceride pyrophosphorylase 2; CDP-diglyceride synthase 2; CDP-diglyceride synthetase 2; CDS 2; CDS2; CTP:phosphatidate cytidylyltransferase 2; D2Wsu127e; Phosphatidate cytidylyltransferase 2 | |
Rabbit | |
Protein G | |
RUO | |
8760 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.995 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O95674 | |
CDS2 | |
Recombinant Human Phosphatidate cytidylyltransferase 2 protein (354-445AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction