Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDTA (Free Acid), ACS Reagent (1,2-Cyclohexylenedinitrilo)tetraacetic Acid Monohydrate), Reagents
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
Color | Colorless/White |
Packaging | Poly Bottle |
Quantity | 500 g |
Specific Gravity | 1.5 |
Grade | ACS |
Physical Form | Solid |
Chemical Name or Material | CDTA (Free Acid), ACS Reagent (1,2-Cyclohexylenedinitrilo)tetraacetic Acid Monohydrate) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction