Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDX1 Monoclonal Antibody, Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 61791

Description
Could play a role in the terminal differentiation of the intestine.Specifications
CDX1 | |
Monoclonal | |
Unconjugated | |
CDX1 | |
caudal type homeo box 1; caudal type homeo box transcription factor 1; caudal type homeobox 1; caudal type homeobox transcription factor 1; Caudal-type homeobox protein 1; caudal-type homeobox protein CDX1; Cdx; Cdx1; Cdx-1; Homeobox protein CDX-1 | |
Mouse | |
Protein A | |
RUO | |
1044 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP assay, Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
P47902 | |
CDX1 | |
This antibody was raised against an N-terminal peptide (a.a. 30-47) of human CDX1 (NP_001795). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title