Learn More
CDX1 Mouse anti-Human, Active Motif™
Mouse Monoclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 61792
Encompass
 View more versions of this product
Catalog No. 50-199-3637
Add to cart
Description
Description
Could play a role in the terminal differentiation of the intestine.
Specifications
Specifications
CDX1
ChIP assay, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
CDX1
Liquid
Mouse
IgG1
10 μg
-20°C, Avoid Freeze/Thaw Cycles
Primary
1044
CDX1
1 mg/mL
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
P47902
caudal type homeo box 1; caudal type homeo box transcription factor 1; caudal type homeobox 1; caudal type homeobox transcription factor 1; Caudal-type homeobox protein 1; caudal-type homeobox protein CDX1; Cdx; Cdx1; Cdx-1; Homeobox protein CDX-1
This antibody was raised against an N-terminal peptide (a.a. 30-47) of human CDX1 (NP_001795).
Protein A
RUO
Antibody
Monoclonal
Human