Learn More
Cdx1 Mouse anti-Human, DyLight 550, Clone: 1B11A9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237338R
Catalog No. NBP237338R
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Cdx1
1B11A9
Western Blot, ELISA
Purified
CDX1
Purified recombinant fragment of human CDX1 (AA: 122-227) expressed in E. Coli.
Protein A or G purified
Chromatin Modifiers, Epigenetics
Primary
1044
Western Blot, ELISA
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human