Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdx1 Rabbit anti-Human, Clone: JG38-50, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275437

Catalog No. NBP275437


Add to cart

Description

Description

Cdx1 Monoclonal antibody specifically detects Cdx1 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Cdx1
JG38-50
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915
Rabbit
IgG
Protein A purified
Chromatin Modifiers, Epigenetics
Primary
1044.0
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
CDX1
Recombinant protein of full lenth sequence of human Cdx1.
28 kDa
100 μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit