Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Mouse anti-Human, Clone: rCDX2/1690, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen CDX2
Clone rCDX2/1690
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122362
View Documents
Novus Biologicals
NBP30744420UG
20 μg
Each for $159.82
Add to cart
 
NB122361
View Documents
Novus Biologicals
NBP30744410UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

CDX2
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Recombinant fragment (around aa150-249) of human CDX2 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
rCDX2/1690
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
1045
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit