Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDX2 Mouse anti-Human, Clone: UMAB265, liquid, UltraMAB™

Mouse Monoclonal Antibody
Supplier: OriGene UM800155

Description
CDX2 is a caudal type homeobox gene that encodes an intestine-specific transcription factor that is expressed early in intestinal development and may be involved in the regulation of proliferation and differentiation of intestinal epithelial cells. It is expressed in the nuclei of epithelial cells throughout the intestine, from duodenum to rectum. The CDX2 protein is expressed in primary and metastatic colorectal carcinomas and has also been demonstrated in the intestinal metaplasia of the stomach and intestinal-type gastric cancer, while it is not expressed in the normal gastric mucosa. Studies have shown that CDX2 is superior marker compared to CK20.Specifications
CDX2 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q99626 | |
CDX2 | |
Full length human recombit protein of human CDX2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin) | |
UMAB265 | |
Unconjugated | |
CDX2 | |
CDX-3, CDX2/AS, CDX3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1045 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title