Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDX2, Mouse anti-Human, DyLight 550, Clone: UPK1A/2923, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275759R
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
CDX2 | |
UPK1A/2923 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDX2 | |
Recombinant full-length human CDX2protein | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
The specificity of this monoclonal antibody to its intended target was validated by HuProt Human Proteome Microarray, containing more than 19,000, full-length human proteins. The intestine-specific transcription factors CDX1 and CDX2 are important for directing intestinal development, differentiation, proliferation and maintenance of the intestinal phenotype. CDX2 protein expression has been seen in GI carcinomas. Anti-CDX2 has been useful to establish GI origin of metastatic adenocarcinomas and carcinoids and is especially useful to distinguish metastatic colorectal adenocarcinoma from lung adenocarcinoma. However, mucinous carcinomas of the ovary also express CDX2 protein. It limits the usefulness of this marker in the distinction of metastatic colorectal adenocarcinoma from mucinous carcinoma of the ovary. |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
DyLight 550 | |
Purified,Conjugated | |
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
Mouse | |
IgG1 | |
Primary | |
0.1 mL | |
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title