Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Mouse anti-Human, FITC, Clone: 3G9B9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237296F

Catalog No. NBP237296F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CDX2
3G9B9
Flow Cytometry, ELISA
Purified
CDX2
Purified recombinant fragment of human CDX2 (AA: 176-303) expressed in E. Coli.
Protein A or G purified
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Primary
1045
Flow Cytometry, ELISA
FITC
PBS with 0.05% Sodium Azide
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit