Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2, Mouse anti-Human, Janelia Fluor 549, Clone: UPK1A/2923, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275759JF549

Catalog No. NB008933


Add to cart

Description

Description

CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

CDX2
UPK1A/2923
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
50mM Sodium Borate with 0.05% Sodium Azide
CDX2
Recombinant full-length human CDX2protein
Monoclonal
Protein A or G purified
RUO
Store at 4°C in the dark.
The specificity of this monoclonal antibody to its intended target was validated by HuProt Human Proteome Microarray, containing more than 19,000, full-length human proteins. The intestine-specific transcription factors CDX1 and CDX2 are important for directing intestinal development, differentiation, proliferation and maintenance of the intestinal phenotype. CDX2 protein expression has been seen in GI carcinomas. Anti-CDX2 has been useful to establish GI origin of metastatic adenocarcinomas and carcinoids and is especially useful to distinguish metastatic colorectal adenocarcinoma from lung adenocarcinoma. However, mucinous carcinomas of the ovary also express CDX2 protein. It limits the usefulness of this marker in the distinction of metastatic colorectal adenocarcinoma from mucinous carcinoma of the ovary.
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin)
Janelia Fluor 549
Purified,Conjugated
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Mouse
IgG1
Primary
0.1 mL
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit