Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Mouse anti-Human, PE-Cyanine7, Clone: CDX2/1690, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254472PECY7

Catalog No. N254472PEC7


Add to cart

Description

Description

CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CDX2
CDX2/1690
Flow Cytometry
Conjugated, Purified
CDX2
Recombinant human CDX2 protein fragment (aa150-249) (exact sequence is proprietary)
Protein A or G purified
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
Primary
1045.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit