Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDYL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530995
Description
Recommended positive controls: NT2D1, U87-MG, mouse kidney. Predicted reactivity: Mouse (84%), Rat (84%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Chromodomain Y is a primate-specific Y-chromosomal gene family expressed exclusively in the testis and implicated in infertility. Although the Y-linked genes are testis-specific, this autosomal gene is ubiquitously expressed. The Y-linked genes arose by retrotransposition of an mRNA from this gene, followed by amplification of the retroposed gene. Proteins encoded by this gene superfamily possess a chromodomain, a motif implicated in chromatin binding and gene suppression, and a catalytic domain believed to be involved in histone acetylation. Multiple proteins are encoded by transcript variants of this gene.
Specifications
CDYL | |
Polyclonal | |
Unconjugated | |
CDYL | |
AI325931; CDYL; CDYL1; CDY-like; CDY-like, autosomal; chromodomain protein Y-like variant 2; chromodomain protein Y-like variant 3; chromodomain protein Y-like variant 4; chromodomain protein, Y chromosome-like; chromodomain protein, Y-like; chromodomain Y like; chromodomain Y-like protein; chromodomain Y-like protein pseudogene; Crotonyl-CoA hydratase; DKFZp586C1622; MGC131936; Putative histone acetyltransferase Cdyl; Putative tubulin acetyltransferase Cdyl; testis-specific chromodomain Y-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12593, 9425 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.54 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9WTK2, Q9Y232 | |
CDYL | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 544 of Human CDYL. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction