Learn More
Aviva Systems Biology Corp CDYL2 Antibody - N-terminal region (ARP49048_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDYL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDYL2. Alternate names: FLJ38866, PCCP1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60003