Tocris™ CE3F4

Uncompetitive Epac1 inhibitor

Manufacturer: Tocris™ 4793/50

 View more versions of this product

Catalog No. 47-935-0

  N/A
Add to Cart

Disclaimers:

For Research Use Only

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Inhibitors CE3F4
    Purity >98
    Molecular Weight 351.01
    Quantity 50mg
    Product Type CE3F4

    Uncompetitive Epac1 inhibitor. Blocks Epac-induced Rap activation and prevents isoprenaline-induced autophagy flux in cardiomyocytes. Has no effect on PKA activity in the presence of cAMP.

    • Chemical Name: 5,7-Dibromo-6-fluoro-3,4-dihydro-2-methyl-1(2H)-quinolinecarboxaldehyde
    • Formula: C11H10Br2FNO
    • CAS Number: 143703-25-7
    • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit