Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer:  Thermo Scientific™ MS613PABX

 View more versions of this product

Catalog No. MS613PABX

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: COL-1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Immunogen: Human colon carcinoma extract

Molecular Weight: 180 kDa

Positive Control: LS174T cells, Colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CEA (Carcinoembryonic Antigen)/CD66e Ab-3
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
COL-1
Unconjugated
Concentrated
Human colon carcinoma extract
200μL
Oncogenesis
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit