Learn More
CEACAM/CD66 Mouse anti-Human, Alexa Fluor™ 594, Clone: COL-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233306AF594
Catalog No. NP233306A94
Add to cart
Description
Description
CEACAM/CD66 Monoclonal antibody specifically detects CEACAM/CD66 in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
CEACAM/CD66
COL-1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CEACAM1
Colon carcinoma cell extract was used as the immunogen for the CEACAM1 antibody.
0.1 mL
Primary
634
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
This antibody recognizes proteins of 80-200kDa, identified as different members of CEACAM family. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEACAM is not found in benign glands, stroma, or malignant prostatic cells