Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Mouse anti-Human, Clone: B3-17, Novus Biologicals™
Mouse Monoclonal Antibody
$212.24 - $551.00
Specifications
Antigen | CEACAM1/CD66a |
---|---|
Clone | B3-17 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA, CyTOF-ready.Specifications
CEACAM1/CD66a | |
LYOPH | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
CEACAM1 | |
IgG1 κ | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
B3-17 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
634 | |
recombinant human soluble CEACAM-1-Fc (produced in HEK293 cells) | |
Primary | |
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. | |
Anti-human CEACAM-1 (B3) interacts solely with human CEACAM-1 and shows no cross-reactivity with other CEACAMs expressed in human or other species, Clone B3 binds to the A1-B-Domain. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title