Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Mouse anti-Rat, Allophycocyanin/Cy7, Clone: 11-1H, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259674APCCY7

Catalog No. N259674APC7


Add to cart

Description

Description

CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

CEACAM1/CD66a
11-1H
Flow Cytometry
Conjugated, Purified
CEACAM1
Recombinant soluble rat CEACAM1-Fc produced in HEK293 cells
Protein A purified
Apoptosis, Cancer
Primary
634.0
Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2. Does not bind to the N-domain of rat CEACAM1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit