Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Mouse anti-Rat, Allophycocyanin/Cy7, Clone: F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259673APCCY7

Catalog No. N259673APC7


Add to cart

Description

Description

CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

CEACAM1/CD66a
F7
Flow Cytometry
Conjugated, Purified
CEACAM1
The immunogen for this antibody is proprietary.
Protein A purified
Apoptosis, Cancer
Primary
634.0
Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Binds specific to the N-domain of rat CEACAM1. Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit