Learn More
CEACAM1 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Manufacturer:  Abnova Corporation H00000634B02P
Catalog No. 89-032-197
Add to cart
Description
Description
Mouse polyclonal antibody raised against a full-length human CEACAM1 protein.
  • High specificity
  • High reactivity
Specifications
Specifications
CEACAM1 purified MaxPab mouse
Unconjugated
In 1x PBS, pH 7.4
Liquid
CEACAM1
CEACAM1 (NP_001020083.1, 1 a.a. ~ 464 a.a) full-length human protein.
RUO
Primary
634
Western Blot
Mouse polyclonal antibody raised against a full-length human CEACAM1 protein.
NM_001024912.1
BGP|BGP1|BGPI
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)