Learn More
Aviva Systems Biology Corp CEACAM19 Antibody - middle region (ARP49488_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49488_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEACAM19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CEACAM19. Alternate names: CEAL1, MGC105097ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60095