Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM19 Mouse anti-Human, Alexa Fluor 532, Clone: Z5P2D9*A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP253124AF532

Catalog No. NP23124A32


Add to cart

Description

Description

CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

CEACAM19
Z5P2D9&asterisk;A9
Western Blot, ELISA
carcinoembryonic antigen-related cell adhesion molecule 19
Mouse
IgG
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CEACAM19
Peptide sequence - RSINPARPLP (BLAST analysis revealed 100% identity with Gorilla)
0.1 mL
Primary
56971
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit