Learn More
CEACAM2 Rat anti-Mouse, Alexa Fluor 647, Clone-935610, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB9214R025UG
Catalog No. FAB9214R025
Add to cart
This item is not returnable. View return policy
Description
Description
CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CEACAM2
935610
Flow Cytometry 0.1 ug/10^6 cells
Q925P2
Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2 Gln35-Asp422 Accession # Q925P2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
26367
Mouse
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
Rat
IgG1
25 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse CEACAM-2 in direct ELISAs. Stains mouse CEACAM-2 transfectants but not irrelevant transfectants in flow cytometry.