Learn More
CEACAM2 Rat anti-Mouse, Alexa Fluor 750, Clone: 935610, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB9214S100UG
Catalog No. FAB9214S100
Add to cart
This item is not returnable. View return policy
Description
Description
CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CEACAM2
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rat
IgG1
RUO
Primary
26367
Mouse
Flow Cytometry
935610
Flow Cytometry 0.25-1 ug/10^6 cells
Q925P2
Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2 Gln35-Asp422 Accession # Q925P2
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse CEACAM-2 in direct ELISAs.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)