Learn More
CEACAM4 Mouse anti-Human, Alexa Fluor 350, Clone: 822608, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7845U100UG
Catalog No. FAB7845U100
Add to cart
This item is not returnable. View return policy
Description
Description
CEACAM4 Monoclonal antibody specifically detects CEACAM4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CEACAM4
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O75871
CEACAM4
E. coli-derived recombinant human CEACAM-4 Phe36-Val140 Accession # O75871
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human CEACAM-4 in direct ELISAs.
Flow Cytometry
822608
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CEACAM4, CGM7, CGM7_HUMAN
Mouse
IgG2b
RUO
Primary
1089
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)