Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Antibody (OTI3D8), DyLight 755, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270407IR

Catalog No. NBP270407IR


Add to Cart

Description

Description

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CEACAM5/CD66e
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CEACAM5
Recombinant protein fragment corresponding to amino acids 35-680 of human CEACAM5/CD66e (NP_004354) produced in SF9 cell.
0.1 mL
Primary
Human
IgG2a
Western Blot, Immunohistochemistry
OTI3D8
Western Blot, Immunohistochemistry
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
Affinity Purified
Apoptosis, Cancer, Cellular Markers, Immunology
1048.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit