Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals CEACAM5/CD66e Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2793435PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM5/CD66e.Specifications
62.5 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM5/CD66e. | |
CEACAM5/CD66e | |
Apoptosis, Cancer, Cellular Markers, Immunology | |
Human | |
CEACAM5 | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.3 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended. |
HRP | |
1048 | |
Antibody Pairs | |
62.5-4000 pg/mL | |
Storage is content dependent. | |
Mouse | |
5 Plates | |
Sandwich ELISA | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title