Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Human anti-Human, DyLight™ 405, Clone: Arcitumomab, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275904V

Catalog No. NBP275904V


Add to cart

Description

Description

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence.
Specifications

Specifications

CEACAM5/CD66e
Arcitumomab
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Conjugated, Purified
CEACAM5
Carcinoembryonic antigen (CEA) describes a set of highly related glycoproteins involved in cell adhesion normally produced in gastrointestinal tissue during fetal development, but only at very low levels in adults.
Protein A purified
Apoptosis, Cancer, Cellular Markers, Immunology
Primary
1048.0
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Human
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Recognises human carcinoembryonic antigen.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit