Learn More
CEACAM5/CD66e, Human anti-Human, PE-Cy5.5, Clone: Arcitumomab, Novus Biologicals™
Human Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP275904PECY55100UL
Catalog No. NB009303
Add to cart
Description
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human Samples. It is validated for Flow Cytometry.
Specifications
Specifications
CEACAM5/CD66e
Arcitumomab
Flow Cytometry
Purified
CEACAM5
Carcinoembryonic antigen (CEA) describes a set of highly related glycoproteins involved in cell adhesion normally produced in gastrointestinal tissue during fetal development, but only at very low levels in adults.
Protein A purified
RUO
Store at 4°C in the dark. Do not freeze.
Monoclonal
Recognises human carcinoembryonic antigen.
Flow Cytometry
PE-Cyanine5.5
PBS
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Human
IgG1 Kappa
100 uL
Apoptosis, Cancer, Cellular Markers, Immunology
Primary
1048
Human